accessibility menu, dialog, popup

UserName

FClClO

CAS RN 65055-17-6

IUPAC Name: 3-chloro-4-fluorobenzoyl chloride
Synonyms: 3-chloro-4-fluorobenzoyl chloride
Molecular Weight (g/mol): 193
Molecular Formula: C7H3Cl2FO
InChi Key: DLZUHSFUBZTBQZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Cl)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results