missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11419676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419676 25 g 25g
11409676 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419676 Supplier Thermo Scientific Alfa Aesar Supplier No. B20217.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65055-17-6
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD00236215
InChI Key DLZUHSFUBZTBQZ-UHFFFAOYSA-N
Synonym 4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f
PubChem CID 2736548
IUPAC Name 3-chloro-4-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1C(=O)Cl)Cl)F

Specifications

Boiling Point 222°C
Refractive Index 1.551
Quantity 25 g
UN Number UN3265
Beilstein 6288904
Sensitivity Moisture sensitive
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-fluorobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.