CAS RN 64118-84-9
IUPAC Name:
2-{2-[(2,6-dichloro-4-hydroxyphenyl)amino]phenyl}acetic acid
Synonyms:
4'-hydroxydiclofenac 4'-OH dcf 4'-hydroxy diclofenac
Molecular Weight (g/mol):
312.15
Molecular Formula:
C14H11Cl2NO3
InChi Key:
KGVXVPRLBMWZLG-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC=CC=C1NC1=C(Cl)C=C(O)C=C1Cl
1
–
4
of
4
results
2
4'-Hydroxydiclofenac, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
4’-Hydroxy Diclofenac, TRC
CAS: 64118-84-9 Molecular Formula: C14 H11 Cl2 N O3 Molecular Weight (g/mol): 312.15 Synonym: Benzeneacetic acid, 2-[(2,6-dichloro-4-hydroxyphenyl)amino]-,2-[(2,6-Dichloro-4-hydroxyphenyl)amino]benzeneacetic acid,4'-Hydroxydiclofenac,Diclofenac-4-hydroxy,4'-Hydroxy Diclofenac IUPAC Name: 2-[2-(2,6-dichloro-4-hydroxyanilino)phenyl]acetic acid SMILES: OC(=O)Cc1ccccc1Nc2c(Cl)cc(O)cc2Cl