CAS RN 638-94-8
CAS RN 638-94-8
IUPAC Name:
(2S,4R,8S,12S)-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Synonyms:
desonide
Molecular Weight (g/mol):
416.51
Molecular Formula:
C24H32O6
InChi Key:
WBGKWQHBNHJJPZ-YWZQBGSISA-N
SMILES:
CC1(C)O[C@@H]2C[C@H]3C4CCC5=CC(=O)C=CC5(C)[C@H]4C(O)CC3(C)[C@@]2(O1)C(=O)CO
1
–
2
of
2
results
1
Thermo Scientific Chemicals Desonide
CAS: 638-94-8 Molecular Formula: C24H32O6
2
16alpha,17-[1-Methylethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione (Desonide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.