accessibility menu, dialog, popup

UserName

CAS RN 62669-70-9

ClH3COONH2OH2N(?)

CAS RN 62669-70-9

IUPAC Name: 3,6-diamino-9-[2-(methoxycarbonyl)phenyl]-10λ⁴-xanthen-10-ylium chloride
Synonyms: rhodamine 123
Molecular Weight (g/mol): 380.83
Molecular Formula: C21H17ClN2O3
InChi Key: TUFFYSFVSYUHPA-UHFFFAOYSA-M
SMILES: [Cl-].COC(=O)C1=CC=CC=C1C1=C2C=CC(N)=CC2=[O+]C2=CC(N)=CC=C12
Color 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)