accessibility menu, dialog, popup

UserName

CAS RN 610769-35-2

BOHOHHONOHOOHONOHOOOHOHOHH2NHO

CAS RN 610769-35-2

IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol; boric acid
Synonyms: boric acid; edetic acid; tris buffer
Molecular Weight (g/mol): 475.21
Molecular Formula: C14H30BN3O14
InChi Key: OSBLTNPMIGYQGY-UHFFFAOYSA-N
SMILES: OB(O)O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Quantity 
  • (2)
  • (1)
pH 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)