accessibility menu, dialog, popup

UserName

NH2IIOHO

CAS RN 609-86-9

IUPAC Name: 2-amino-3,5-diiodobenzoic acid
Synonyms: 2-amino-3,5-diiodobenzoic acid
Molecular Weight (g/mol): 388.93
Molecular Formula: C7H5I2NO2
InChi Key: RFIBDMCPIREZKC-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(I)C=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results