accessibility menu, dialog, popup

UserName

CAS RN 597-64-8

CH3SnH3CH3CH3C

CAS RN 597-64-8

IUPAC Name: tetraethylstannane
Synonyms: tetraethyltin tetraethyl tin
Molecular Weight (g/mol): 234.96
Molecular Formula: C8H20Sn
InChi Key: RWWNQEOPUOCKGR-UHFFFAOYSA-N
SMILES: CC[Sn](CC)(CC)CC
Boiling Point 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)