accessibility menu, dialog, popup

UserName

CH3OClF

CAS RN 59189-98-9

IUPAC Name: 2-fluoro-4-methylbenzoyl chloride
Synonyms: 2-fluoro-4-methylbenzoyl chloride
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: SVSVOPWCUHQFAM-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results