missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylbenzoyl chloride, 97%, Thermo Scientific™

Product Code. 15494057
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494057 1 g 1g
15404067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494057 Supplier Thermo Scientific Alfa Aesar Supplier No. H31292.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59189-98-9
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD04115879
InChI Key SVSVOPWCUHQFAM-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-fluoro-4-methyl-9ci, 2-fluoro-4-methyl-benzoyl chloride, benzoyl chloride, 2-fluoro-4-methyl
PubChem CID 5173367
IUPAC Name 2-fluoro-4-methylbenzoyl chloride
SMILES CC1=CC(F)=C(C=C1)C(Cl)=O

Specifications

Melting Point 43°C to 47°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methylbenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.