accessibility menu, dialog, popup

UserName

CH3NCH3CH3H3COO

CAS RN 59138-84-0

IUPAC Name: tetramethylazanium formate
Synonyms: tetramethylammonium ion formate
Molecular Weight (g/mol): 119.16
Molecular Formula: C5H13NO2
InChi Key: WWIYWFVQZQOECA-UHFFFAOYSA-M
SMILES: [O-]C=O.C[N+](C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results