CAS RN 58368-67-5
CAS RN 58368-67-5
IUPAC Name:
pentan-3-yl formate
Synonyms:
pentan-3-yl formate
Molecular Weight (g/mol):
116.16
Molecular Formula:
C6H12O2
InChi Key:
YMJOAYHERILFIM-UHFFFAOYSA-N
SMILES:
CCC(CC)OC=O
1
–
1
of
1
results
1
3-Pentyl formate, 94%, Thermo Scientific™
CAS: 58368-67-5 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 MDL Number: MFCD00049220 InChI Key: YMJOAYHERILFIM-UHFFFAOYSA-N Synonym: 3-pentyl formate,acmc-1b22e,formic acid 1-ethylpropyl ester PubChem CID: 13878880 IUPAC Name: pentan-3-yl formate SMILES: CCC(CC)OC=O