CAS RN 56828-40-1
CAS RN 56828-40-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Minoxidil Beta-D-Glucuronide, TRC
CAS: 56828-40-1 Molecular Formula: C15 H23 N5 O7 Molecular Weight (g/mol): 385.37 Synonym: β-D-Glucopyranuronic acid, 1-[[4-amino-2-imino-6-(1-piperidinyl)-1(2H)-pyrimidinyl]oxy]-1-deoxy- (9CI),Minoxidil β-D-Glucuronide IUPAC Name: (2S,3S,4S,5R,6S)-6-(2,6-diamino-4-piperidin-1-ylpyrimidin-1-ium-1-yl)oxy-3,4,5-trihydroxyoxane-2-carboxylate SMILES: Nc1cc(nc(N)[n+]1O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)[O-])N3CCCCC3