missing translation for 'onlineSavingsMsg'
Learn More

Minoxidil Beta-D-Glucuronide, TRC

Product Code. 30479960
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479960 5 mg 5mg
30472521 1 mg 1mg
30469749 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479960 Supplier Toronto Research Chemicals Supplier No. TRCM3450505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56828-40-1
Molecular Formula C15 H23 N5 O7
Molecular Weight (g/mol) 385.37
Synonym β-D-Glucopyranuronic acid, 1-[[4-amino-2-imino-6-(1-piperidinyl)-1(2H)-pyrimidinyl]oxy]-1-deoxy- (9CI), Minoxidil β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-(2,6-diamino-4-piperidin-1-ylpyrimidin-1-ium-1-yl)oxy-3,4,5-trihydroxyoxane-2-carboxylate
SMILES Nc1cc(nc(N)[n+]1O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)[O-])N3CCCCC3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H23N5O7/c16-7-6-8(19-4-2-1-3-5-19)18-15(17)20(7)27-14-11(23)9(21)10(22)12(26-14)13(24)25/h6,9-12,14,21-23H,1-5H2,(H4,16,17,18,24,25)/t9-,10-,11+,12-,14-/m0/s1
Formula Weight 385.1597
Chemical Name or Material Minoxidil Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.