CAS RN 56794-28-6
IUPAC Name:
4-chloro-N,N-diethylbutanamide
Synonyms:
4-chloro-N,N-diethylbutanamide
Molecular Weight (g/mol):
177.67
Molecular Formula:
C8H16ClNO
InChi Key:
HHBZYNJHOOGYPZ-UHFFFAOYSA-N
SMILES:
CCN(CC)C(=O)CCCCl
1
–
1
of
1
results
1
4-Chloro-N,N-diethylbutyramide, 97%, Thermo Scientific™
CAS: 56794-28-6 Molecular Formula: C8H16ClNO Molecular Weight (g/mol): 177.672 MDL Number: MFCD03988556 InChI Key: HHBZYNJHOOGYPZ-UHFFFAOYSA-N Synonym: 4-chloro-n,n-diethyl-butyramide,4-chloro-n,n-diethylbutyramide PubChem CID: 2434869 IUPAC Name: 4-chloro-N,N-diethylbutanamide SMILES: CCN(CC)C(=O)CCCCl