accessibility menu, dialog, popup

UserName

CAS RN 563-83-7

H3CCH3NH2O

CAS RN 563-83-7

IUPAC Name: 2-methylpropanamide
Synonyms: isobutyramide
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: WFKAJVHLWXSISD-UHFFFAOYSA-N
SMILES: CC(C)C(N)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results