accessibility menu, dialog, popup

UserName

CAS RN 5600-21-5

CH3ClNH2NN

CAS RN 5600-21-5

IUPAC Name: 4-chloro-6-methylpyrimidin-2-amine
Synonyms: 4-chloro-6-methylpyrimidin-2-amine
Molecular Weight (g/mol): 143.57
Molecular Formula: C5H6ClN3
InChi Key: NPTGVVKPLWFPPX-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC(N)=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)