Learn More
2-Amino-4-chloro-6-methylpyrimidine, TRC
Product Code. 30323261
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323261 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323261 Supplier Toronto Research Chemicals Supplier No. TRCA617418100MG
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-4-chloro-6-methylpyrimidine
CAS 5600-21-5
Molecular Formula C5 H6 Cl N3
Quantity 100 mg
Synonym Pyrimidine, 2-amino-4-chloro-6-methyl- (6CI,7CI,8CI), 4-Chloro-6-methyl-2-pyrimidinamine, 2-Amino-4-chloro-6-methylpyrimidine, 2-Amino-6-chloro-4-methylpyrimidine, 4-Chloro-6-methylpyrimidin-2-ylamine, AM, AM (nitrification inhibitor), NSC 23661, NSC 7892
InChI Formula InChI=1S/C5H6ClN3/c1-3-2-4(6)9-5(7)8-3/h2H,1H3,(H2,7,8,9)
SMILES Cc1cc(Cl)nc(N)n1
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
Molecular Weight (g/mol) 143.57
Formula Weight 143.025