Learn More
2-Amino-4-chloro-6-methylpyrimidine, TRC
Product Code. 30310746
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310746 Supplier Toronto Research Chemicals Supplier No. TRCA6174181G
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-4-chloro-6-methylpyrimidine
CAS 5600-21-5
Molecular Formula C5 H6 Cl N3
Quantity 1 g
Synonym Pyrimidine, 2-amino-4-chloro-6-methyl- (6CI,7CI,8CI), 4-Chloro-6-methyl-2-pyrimidinamine, 2-Amino-4-chloro-6-methylpyrimidine, 2-Amino-6-chloro-4-methylpyrimidine, 4-Chloro-6-methylpyrimidin-2-ylamine, AM, AM (nitrification inhibitor), NSC 23661, NSC 7892
InChI Formula InChI=1S/C5H6ClN3/c1-3-2-4(6)9-5(7)8-3/h2H,1H3,(H2,7,8,9)
SMILES Cc1cc(Cl)nc(N)n1
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
Molecular Weight (g/mol) 143.57
Formula Weight 143.025