CAS RN 55804-66-5
CAS RN 55804-66-5
IUPAC Name:
ethyl 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Synonyms:
coumarin 314 coumarin 504
Molecular Weight (g/mol):
313.35
Molecular Formula:
C18H19NO4
InChi Key:
VMJKUPWQKZFFCX-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
1
–
1
of
1
results
1
Coumarin 314, 99%, pure, laser grade
CAS: 55804-66-5 Molecular Formula: C18H19NO4 Molecular Weight (g/mol): 313.353 MDL Number: MFCD00051334 InChI Key: VMJKUPWQKZFFCX-UHFFFAOYSA-N Synonym: 1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one PubChem CID: 72653 ChEBI: CHEBI:51940 SMILES: CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3