accessibility menu, dialog, popup

UserName

CH3ClH2C

CAS RN 557-98-2

IUPAC Name: 2-chloroprop-1-ene
Synonyms: 2-chloropropene
Molecular Weight (g/mol): 76.52
Molecular Formula: C3H5Cl
InChi Key: PNLQPWWBHXMFCA-UHFFFAOYSA-N
SMILES: CC(Cl)=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results