CAS RN 55079-83-9
CAS RN 55079-83-9
IUPAC Name:
(2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
Synonyms:
acitretin acitretin - chemical acitretinum andreu brand of acitretin 13-cis-acitretin soriatane acitretin, (z,e,e,e)-isomer all-trans-acitretin (z,e,e,e)-acitretin 13-cis-etretin
Molecular Weight (g/mol):
326.44
Molecular Formula:
C21H26O3
InChi Key:
IHUNBGSDBOWDMA-AQFIFDHZSA-N
SMILES:
COC1=CC(C)=C(\C=C\C(\C)=C\C=C\C(\C)=C\C(O)=O)C(C)=C1C
1
–
3
of
3
results
1
Thermo Scientific Chemicals Acitretin
CAS: 55079-83-9 Molecular Formula: C21H26O3
2
Acitretin, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Acitretin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.