accessibility menu, dialog, popup

UserName

H3CCH3OOOCH3(E/Z)

CAS RN 5466-77-3

IUPAC Name: 2-ethylhexyl (2E)-3-(4-methoxyphenyl)prop-2-enoate
Synonyms: parsol ethylhexyl methoxycinnamate 2-ethylhexyl methoxycinnamate octyl methoxycinnamate octinoxate parsol mox
Molecular Weight (g/mol): 290.4
Molecular Formula: C18H26O3
InChi Key: YBGZDTIWKVFICR-JLHYYAGUNA-N
SMILES: CCCCC(CC)COC(=O)\C=C\C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results