CAS RN 5428-61-5
CAS RN 5428-61-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N4-(5-Chloro-4-quinolinyl)-N1,N1-diethyl-1,4-pentanediamine, TRC
CAS: 5428-61-5 Molecular Formula: C18H26ClN3 Molecular Weight (g/mol): 319.87 Synonym: 5-Chloro-4-[[4-(diethylamino)-1-methylbutyl]amino]quinoline IUPAC Name: 4-N-(5-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine SMILES: CCN(CC)CCCC(C)Nc1ccnc2cccc(Cl)c12