missing translation for 'onlineSavingsMsg'
Learn More

N4-(5-Chloro-4-quinolinyl)-N1,N1-diethyl-1,4-pentanediamine, TRC

Product Code. 30364009
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364009 10 mg 10mg
30355106 50 mg 50mg
30354324 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364009 Supplier Toronto Research Chemicals Supplier No. TRCC54261510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5428-61-5
Molecular Formula C18H26ClN3
Molecular Weight (g/mol) 319.87
Synonym 5-Chloro-4-[[4-(diethylamino)-1-methylbutyl]amino]quinoline
IUPAC Name 4-N-(5-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine
SMILES CCN(CC)CCCC(C)Nc1ccnc2cccc(Cl)c12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H26ClN3/c1-4-22(5-2)13-7-8-14(3)21-17-11-12-20-16-10-6-9-15(19)18(16)17/h6,9-12,14H,4-5,7-8,13H2,1-3H3,(H,20,21)
Formula Weight 319.1815
Chemical Name or Material N4-(5-Chloro-4-quinolinyl)-N1,N1-diethyl-1,4-pentanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.