CAS RN 54063-54-6
CAS RN 54063-54-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Reproterol, TRC
CAS: 54063-54-6 Molecular Formula: C18 H23 N5 O5 Molecular Weight (g/mol): 389.41 Synonym: 7-[3-[[2-(3,5-Dihydroxyphenyl)-2-hydroxyethyl]amino]propyl]-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione,D 1959; IUPAC Name: 7-[3-[[2-(3,5-dihydroxyphenyl)-2-hydroxyethyl]amino]propyl]-1,3-dimethylpurine-2,6-dione SMILES: O=C1C2=C(N=CN2CCCNCC(O)C=3C=C(O)C=C(O)C3)N(C(=O)N1C)C