missing translation for 'onlineSavingsMsg'
Learn More

Reproterol, TRC

Product Code. 17999844
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17999844 100 mg 100mg
17959454 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17999844 Supplier Toronto Research Chemicals Supplier No. TRCR144650100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54063-54-6
Molecular Formula C18 H23 N5 O5
Molecular Weight (g/mol) 389.41
Synonym 7-[3-[[2-(3,5-Dihydroxyphenyl)-2-hydroxyethyl]amino]propyl]-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione, D 1959;
IUPAC Name 7-[3-[[2-(3,5-dihydroxyphenyl)-2-hydroxyethyl]amino]propyl]-1,3-dimethylpurine-2,6-dione
SMILES O=C1C2=C(N=CN2CCCNCC(O)C=3C=C(O)C=C(O)C3)N(C(=O)N1C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H23N5O5/c1-21-16-15(17(27)22(2)18(21)28)23(10-20-16)5-3-4-19-9-14(26)11-6-12(24)8-13(25)7-11/h6-8,10,14,19,24-26H,3-5,9H2,1-2H3
Formula Weight 389.1699
Chemical Name or Material Reproterol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.