accessibility menu, dialog, popup

UserName

CAS RN 53957-33-8

CH3CH3HO

CAS RN 53957-33-8

IUPAC Name: (2,5-dimethylphenyl)methanol
Synonyms: (2,5-dimethylphenyl)methanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: LEBQTCCCNMTXSF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(CO)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results