accessibility menu, dialog, popup

UserName

CAS RN 518070-19-4

IUPAC Name: 3-fluoro-5-methylbenzoic acid
Synonyms: 3-fluoro-5-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: XPUFVYIUPYNLPD-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results