Learn More
3-Fluoro-5-methylbenzoic acid, 97%, Thermo Scientific™
Product Code. 11392599
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392599 5 g 5g
11382599 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11392599 Supplier Thermo Scientific Alfa Aesar Supplier No. H26677.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Intermediate in organic syntheses.
Solubility
Soluble in water (25 deg C)
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 518070-19-4
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD03094313
InChI Key XPUFVYIUPYNLPD-UHFFFAOYSA-N
Synonym 3-fluoro-5-methylbenzoicacid, 3-fluoro-5-methyl-benzoic acid, benzoic acid, 3-fluoro-5-methyl, pubchem1396, 3-carboxy-5-fluorotoluene, acmc-1b05f, ksc498a8l, 3-fluoro-5-methybenzoic acid, rarechem al bo 2238, 3-fluoro-5-methyl benzoic acid
PubChem CID 2778447
IUPAC Name 3-fluoro-5-methylbenzoic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)F
Specifications
Melting Point 141°C to 143°C
Quantity 5 g
Solubility Information Soluble in water (25 deg C)
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-methylbenzoic acid