accessibility menu, dialog, popup

UserName

CAS RN 505084-55-9

NClFFFI

CAS RN 505084-55-9

IUPAC Name: 2-chloro-4-iodo-5-(trifluoromethyl)pyridine
Synonyms: 2-chloro-4-iodo-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 307.44
Molecular Formula: C6H2ClF3IN
InChi Key: XEEQBBXEZSNIQV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(Cl)C=C1I
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)