accessibility menu, dialog, popup

UserName

H3COFFFOCH3

CAS RN 503464-99-1

IUPAC Name: 1-[2-methoxy-5-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[2-methoxy-5-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: FPJNPSGNLVIBBS-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)C(F)(F)F)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results