missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxy-5'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 15455397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455397 5 g 5g
15445397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455397 Supplier Thermo Scientific Alfa Aesar Supplier No. H32652.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
In synthesis and evaluation of disubstituted N1- and N3-Imidazolidin-2-ones Acting as Potential Immunosuppressive Agents

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503464-99-1
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.18
MDL Number MFCD09832300
InChI Key FPJNPSGNLVIBBS-UHFFFAOYSA-N
Synonym 1-2-methoxy-5-trifluoromethylphenyl ethanone, 1-2-methoxy-5-trifluoromethyl phenyl ethanone, 2'-methoxy-5'-trifluoromethyl acetophenone, ethanone, 1-2-methoxy-5-trifluoromethyl phenyl, 2-methoxy-5-trifluoromethyl acetophenone, 2'-methoxy-5'-trifluoromethyl-acetophenone, 1-acetyl-2-methoxy-5-trifluoromethyl benzene, 1-2-methoxy-5-trifluoromethyl phenyl ethan-1-one
PubChem CID 9942601
IUPAC Name 1-[2-methoxy-5-(trifluoromethyl)phenyl]ethanone
SMILES COC1=C(C=C(C=C1)C(F)(F)F)C(C)=O

Specifications

Melting Point 45°C to 47°C
Quantity 5 g
Solubility Information Slightly soluble.
Formula Weight 218.17
Percent Purity 97%
Chemical Name or Material 2'-Methoxy-5'-(trifluoromethyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.