CAS RN 43203-35-6
CAS RN 43203-35-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,9-Dichloro-3,7-diazanonane Dihydrochloride, TRC
CAS: 43203-35-6 Molecular Formula: C7 H16 Cl2 N2 Molecular Weight (g/mol): 199.121 Synonym: NSC 70806,N,N'-Bis(2-chloroethyl)-1,3-propanediamine Dihydrochloride,N1,N3-Bis(2-chloroethyl)-1,3-propanediamine Hydrochloride IUPAC Name: N,N'-bis(2-chloroethyl)propane-1,3-diamine SMILES: ClCCNCCCNCCCl