missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dichloro-3,7-diazanonane Dihydrochloride, TRC

Product Code. 30379386
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379386 25 mg -
30377070 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379386 Supplier Toronto Research Chemicals Supplier No. TRCD43404025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43203-35-6
Molecular Formula C7 H16 Cl2 N2
Molecular Weight (g/mol) 199.121
Synonym NSC 70806, N,N'-Bis(2-chloroethyl)-1,3-propanediamine Dihydrochloride, N1,N3-Bis(2-chloroethyl)-1,3-propanediamine Hydrochloride
IUPAC Name N,N'-bis(2-chloroethyl)propane-1,3-diamine
SMILES ClCCNCCCNCCCl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H16Cl2N2/c8-2-6-10-4-1-5-11-7-3-9/h10-11H,1-7H2
Formula Weight 198.069
Chemical Name or Material 1,9-Dichloro-3,7-diazanonane Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.