accessibility menu, dialog, popup

UserName

CAS RN 423768-47-2

CH3H3CSOOSSOOClOOH3C

CAS RN 423768-47-2

IUPAC Name: methyl 3-(chlorosulfonyl)-4-(propane-2-sulfonyl)thiophene-2-carboxylate
Synonyms: methyl 3-(chlorosulfonyl)-4-(propane-2-sulfonyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 346.81
Molecular Formula: C9H11ClO6S3
InChi Key: FHFIRKJBLQUUAD-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C(=CS1)S(=O)(=O)C(C)C)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results