accessibility menu, dialog, popup

UserName

ONOFFFF

CAS RN 393-09-9

IUPAC Name: 4-fluoro-1-nitro-2-(trifluoromethyl)benzene
Synonyms: 4-fluoro-1-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 209.1
Molecular Formula: C7H3F4NO2
InChi Key: WMQOSURXFLBTPC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C=C1C(F)(F)F
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results