Learn More
5-Fluoro-2-nitrobenzotrifluoride, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 5-Fluoro-2-nitrobenzotrifluoride |
| CAS | 393-09-9 |
| Molecular Formula | C7H3F4NO2 |
| Quantity | 5 g |
| Synonym | 4-Fluoro-1-nitro-2-(trifluoromethyl)benzene, alpha,alpha,alpha,5-Tetrafluoro-2-nitrotoluene, 1-Fluoro-3-trifluoromethyl-4-nitrobenzene, 4-Fluoro-2-(trifluoromethyl)nitrobenzene, 4-Fluoro-6-(trifluoromethyl)nitrobenzene, 4-Nitro-3-(trifluoromethyl)fluorobenzene, 5-Fluoro-2-nitro(trifluoromethyl)benzene, NSC 10304 |
| InChI Formula | InChI=1S/C7H3F4NO2/c8-4-1-2-6(12(13)14)5(3-4)7(9,10)11/h1-3H |
| SMILES | [O-][N+](=O)c1ccc(F)cc1C(F)(F)F |
| IUPAC Name | 4-fluoro-1-nitro-2-(trifluoromethyl)benzene |
| Molecular Weight (g/mol) | 209.1 |
| Formula Weight | 209.01 |