CAS RN 37988-18-4
CAS RN 37988-18-4
IUPAC Name:
N1,N3,N5-tris(2-hydroxyethyl)benzene-1,3,5-tricarboxamide
Synonyms:
N1,N3,N5-tris(2-hydroxyethyl)benzene-1,3,5-tricarboxamide
Molecular Weight (g/mol):
339.35
Molecular Formula:
C15H21N3O6
InChi Key:
RGWJKANXFYJKHN-UHFFFAOYSA-N
SMILES:
OCCNC(=O)C1=CC(=CC(=C1)C(=O)NCCO)C(=O)NCCO
1
–
2
of
2
results
1
2