missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LM 22A4
Description
LM 22A4 is a potent tropomyosin-related kinase B (TrkB) agonist. Induces activation of Trk, Akt and ERK in mouse hippocampus and striatum and exhibits neurotrophic activity. Reverses deficits in motor task learning in mice following traumatic brain injury; restores respiratory function in a rat model of Rett syndrome.
Specifications
Specifications
| Chemical Name or Material | N,N',N''Tris(2-hydroxyethyl)-1,3,5-benzenetricarboxamide |
| CAS | 37988-18-4 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C15H21N3O6 |
| Formula Weight | Observed MW: 339.34 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?