CAS RN 37743-18-3
CAS RN 37743-18-3
IUPAC Name:
N,N-dimethyl-3,3-diphenyloxolan-2-iminium bromide
Synonyms:
N,N-dimethyl-3,3-diphenyloxolan-2-iminium bromide
Molecular Weight (g/mol):
346.27
Molecular Formula:
C18H20BrNO
InChi Key:
QJPXLCQSAFQCBD-UHFFFAOYSA-M
SMILES:
[Br-].C[N+](C)=C1OCCC1(C1=CC=CC=C1)C1=CC=CC=C1
1
–
4
of
4
results
1
Dimethyl(tetrahydro-3,3-diphenyl-2-furylidene)ammonium Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dimethyl-(tetrahydro-3,3-diphenyl-2-furilidine)ammonium Bromide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Dimethyl(tetrahydro-3,3-diphenyl-2-furylidene)ammonium Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Dimethyl(tetrahydro-3,3-diphenyl-2-furylidene)ammonium Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.