accessibility menu, dialog, popup

UserName

CAS RN 3766-81-2

CH3H3COOHNH3C

CAS RN 3766-81-2

IUPAC Name: 2-(butan-2-yl)phenyl N-methylcarbamate
Synonyms: bassa barizon hopcin baycarb carvil fenobcarb fenobucarb osbac geocarb 50EC BPMC
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: DIRFUJHNVNOBMY-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=CC=C1OC(=O)NC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results