accessibility menu, dialog, popup

UserName

CAS RN 374064-02-5

NNNHOHO

CAS RN 374064-02-5

IUPAC Name: 3-(pyridin-2-yl)-1H-pyrazole-5-carboxylic acid
Synonyms: 5-(pyridin-2-yl)-2H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C9H7N3O2
InChi Key: SJBWHTBPIJXUFP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NN1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results