accessibility menu, dialog, popup

UserName

CAS RN 37248-47-8

HO(R)(S)NH(S)HO(R)HO(S)OH(S)OH(S)OH(R)OH(R)O(R)O(R)HO(S)OH(S)OH(R)OH

CAS RN 37248-47-8

IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(1R,2R,3S,4S,6R)-2,3-dihydroxy-6-(hydroxymethyl)-4-{[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino}cyclohexyl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: validamycin A valimon validacin jinggangmycin
Molecular Weight (g/mol): 497.49
Molecular Formula: C20H35NO13
InChi Key: JARYYMUOCXVXNK-CSLFJTBJSA-N
SMILES: OC[C@H]1C[C@H](N[C@H]2C=C(CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)