Learn More
Validamycin A Sulfate salt (Technical grade), TRC
Specifications
Specifications
| Chemical Name or Material | Validamycin A Sulfate Salt |
| CAS | 37248-47-8 |
| Molecular Formula | C20 H35 N O13 . x[H2SO4] |
| Quantity | 10 mg |
| Synonym | Validamycin A Sulfate, Validamycin A Sulphate Salt |
| InChI Formula | InChI=1S/C20H35NO13.H2O4S/c22-3-6-1-8(12(26)15(29)11(6)25)21-9-2-7(4-23)19(17(31)13(9)27)34-20-18(32)16(30)14(28)10(5-24)33-20;1-5(2,3)4/h1,7-32H,2-5H2;(H2,1,2,3,4)/t7-,8+,9+,10-,11-,12+,13+,14-,15+,16+,17-,18-,19-,20+;/m1./s1 |
| SMILES | OCC1=CC(C(C(C1O)O)O)NC2CC(C(C(C2O)O)OC3OC(C(C(C3O)O)O)CO)CO.[xH2SO4] |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[(1R,2R,3S,4S,6R)-2,3-dihydroxy-6-(hydroxymethyl)-4-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]cyclohexoxy]-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol;sulfuric acid |
| Molecular Weight (g/mol) | 595.56928 |
| Formula Weight | 595.1782 |