CAS RN 37091-65-9
CAS RN 37091-65-9
IUPAC Name:
sodium (2R,5S,6S)-3,3-dimethyl-7-oxo-6-[(2R)-2-[(2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetamido]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms:
sodium (2R,5S,6S)-3,3-dimethyl-7-oxo-6-[(2R)-2-(2-oxoimidazolidine-1-carbonylamino)-2-phenylacetamido]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol):
483.47
Molecular Formula:
C20H22N5NaO6S
InChi Key:
UVOCNBWUHNCKJM-QXKADHPISA-M
SMILES:
[Na+].CC1(C)S[C@H]2[C@@H](NC(=O)[C@H](NC(=O)N3CCNC3=O)C3=CC=CC=C3)C(=O)N2[C@@H]1C([O-])=O
1
–
3
of
3
results
1
Azlocillin Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Azlocillin Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Azlocillin Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.