missing translation for 'onlineSavingsMsg'
Learn More

Azlocillin Sodium Salt, TRC

Product Code. 30330629
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330629 Supplier Toronto Research Chemicals Supplier No. TRCA9300001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azlocillin Sodium Salt
CAS 37091-65-9
Molecular Formula C20H23N5O6S(Na)
Quantity 1 g
Synonym (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[(2R)-2-[[(2-oxo-1-imidazolidinyl)carbonyl]amino]-2-phenylacetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Sodium Salt, [2S-[2α,5α,6β(S*)]]-3,3-Dimethyl-7-oxo-6-[[[[(2-oxo-1-imidazolidinyl)carbonyl]amino]phenylacetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Monosodium Salt, Azlin, Azlocillin Sodium, Monosodium Azlocillin, Securopen, Sodium azlocillin
InChI Formula InChI=1S/C20H23N5O6S/c1-20(2)13(17(28)29)25-15(27)12(16(25)32-20)22-14(26)11(10-6-4-3-5-7-10)23-19(31)24-9-8-21-18(24)30/h3-7,11-13,16H,8-9H2,1-2H3,(H,21,30)(H,22,26)(H,23,31)(H,28,29)/t11-,12-,13+,16-/m1/s1
SMILES CC1([C@@H](N2C([C@@H](N=C([O-])[C@@H](NC(N3CCNC3=O)=O)C4=CC=CC=C4)[C@H]2S1)=O)C(O)=O)C.[Na+]
IUPAC Name (2S,5R,6R)-3,3-dimethyl-7-oxo-6-[[(2R)-2-[(2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 461.492299
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.