missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlocillin Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Azlocillin Sodium Salt |
| CAS | 37091-65-9 |
| Molecular Formula | C20H23N5O6S(Na) |
| Quantity | 250 mg |
| Synonym | (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[(2R)-2-[[(2-oxo-1-imidazolidinyl)carbonyl]amino]-2-phenylacetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Sodium Salt, [2S-[2α,5α,6β(S*)]]-3,3-Dimethyl-7-oxo-6-[[[[(2-oxo-1-imidazolidinyl)carbonyl]amino]phenylacetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Monosodium Salt, Azlin, Azlocillin Sodium, Monosodium Azlocillin, Securopen, Sodium azlocillin |
| InChI Formula | InChI=1S/C20H23N5O6S/c1-20(2)13(17(28)29)25-15(27)12(16(25)32-20)22-14(26)11(10-6-4-3-5-7-10)23-19(31)24-9-8-21-18(24)30/h3-7,11-13,16H,8-9H2,1-2H3,(H,21,30)(H,22,26)(H,23,31)(H,28,29)/t11-,12-,13+,16-/m1/s1 |
| SMILES | CC1([C@@H](N2C([C@@H](N=C([O-])[C@@H](NC(N3CCNC3=O)=O)C4=CC=CC=C4)[C@H]2S1)=O)C(O)=O)C.[Na+] |
| IUPAC Name | (2S,5R,6R)-3,3-dimethyl-7-oxo-6-[[(2R)-2-[(2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Molecular Weight (g/mol) | 461.492299 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?