CAS RN 37055-51-9
CAS RN 37055-51-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Diphenylamine-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diphenylamine-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Diphenyl-d10-amine, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
4
Diphenylamine-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.