missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine-d10, TRC

Product Code. 30371612
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371612 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30371612 Supplier Toronto Research Chemicals Supplier No. TRCD478942500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylamine-d10
CAS 37055-51-9
Purity Grade Notes GC
Molecular Formula C12 2H10 H N
Quantity 500 mg
Synonym Diphenylamine-d10, Diphenylamine D10, Benzen-2,3,4,5,6-d5-amine, N-(phenyl-2,3,4,5,6-d5)-, Benzen-d5-amine, N-(phenyl-d5)- (9CI), N-(Phenyl-2,3,4,5,6-d5)benzen-2,3,4,5,6-d5-amine, Di(perdeuterophenyl)amine
InChI Formula InChI=1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMILES [2H]c1c([2H])c([2H])c(Nc2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]
IUPAC Name 2,3,4,5,6-pentadeuterio-N-(2,3,4,5,6-pentadeuteriophenyl)aniline
Molecular Weight (g/mol) 179.28
Formula Weight 179.1519
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.